Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOR-1C Rabbit anti-Mouse, Rat, Polyclonal, Neuromics™

Rabbit Polyclonal Antibody

Manufacturer:  Neuromics, IncSupplier Diversity Partner RA2000150UL

Encompass

 View more versions of this product

Catalog No. 50-210-0118


Add to cart

Description

Description

MOR-1C Polyclonal antibody specifically detects MOR-1C in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Specifications

Specifications

MOR-1C
Polyclonal
Unconjugated
OPRM1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Conc. Not Determined
whole serum with 0.05% sodium azide
P33535, P42866
BOS_9964; hMOP; KIAA0403; LMOR; MOP; MOP receptor; MOP-R; Mor; MOR1; M-OR-1; MOR-1; MOR-1C; MOR-1O; MORA; Mu opiate receptor; Mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; mu-opioid receptor; muOR; MUOR1; Mu-type opioid receptor; Opioid mu receptor; Opioid Receptor; Opioid receptor B; opioid receptor mu 1; opioid receptor, mu 1; Oprm; OPRM1; Oprrm1; OTTHUMP00000017463; Ror-b
KSCMDRGMRNLLPDDGPRQE
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18390, 25601
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit