Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOR174-9 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Supplier: Novus Biologicals NB110750430.1MG
Description
MOR174-9 Polyclonal specifically detects MOR174-9 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MOR174-9 | |
Polyclonal | |
Unconjugated | |
PBS with No Preservative | |
mOR-EG., Odorant receptor, Olfactory receptor MOR174-9, Olfactory receptor Olfr73, Olfr73, ORL828, ORL828. GENE NAME: Olfr73 | |
A synthetic peptide as a part of mouse olfactory receptor family 174, subfamily 9 (MOR174-9) conjugated to immunogenic carrier protein was used as the immunogen. | |
0.1 mg | |
Olfactory | |
MOR 174-9. | |
Mouse, Rat | |
Purified |
Immunohistochemistry (Frozen), Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
LYOPH | |
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Q920P2 | |
Chicken | |
IgG purified | |
RUO | |
Primary | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction