Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOR174-9 Chicken anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Chicken Polyclonal Antibody
$153.31 - $436.00
Specifications
Antigen | MOR174-9 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11075043
|
Novus Biologicals
NB11075043 |
0.5 mg |
Each for $436.00
|
|
NB018025
|
Novus Biologicals
NB110750430.1MG |
0.1 mg |
Each for $153.31
|
|
Description
MOR174-9 Polyclonal specifically detects MOR174-9 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MOR174-9 | |
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat | |
Q920P2 | |
A synthetic peptide as a part of mouse olfactory receptor family 174, subfamily 9 (MOR174-9) conjugated to immunogenic carrier protein was used as the immunogen. | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Chicken | |
Olfactory | |
PBS with No Preservative | |
mOR-EG., Odorant receptor, Olfactory receptor MOR174-9, Olfactory receptor Olfr73, Olfr73, ORL828, ORL828. GENE NAME: Olfr73 | |
Primary | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
MOR 174-9. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title