Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOR42-3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$153.04 - $436.00
Specifications
Antigen | MOR42-3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11075065
|
Novus Biologicals
NB11075065 |
0.5 mg |
Each for $436.00
|
|
NB018027
|
Novus Biologicals
NB110750650.1MG |
0.1 mg |
Each for $153.04
|
|
Description
MOR42-3 Polyclonal specifically detects MOR42-3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MOR42-3 | |
Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
Odorant receptor S6, Olfactory receptor 544, Olfactory receptor Olfr544, Olfr544., ORL463antibody, ORL471, S79 | |
IgG | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Mouse | |
Q9WU88 | |
A synthetic peptide as a part of mouse olfactory receptor family 42, subfamily 3 (MOR42-3) conjugated to immunogenic carrier protein was used as the immunogen. | |
Primary | |
Reconstitute with 0.5 ml sterilized water. Add glycerol (1:1) for additional stability. | |
MOR42-3. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title