Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mordant Yellow 10, pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6054-99-5
C13H8N2Na2O6S
366.255
CWQLKNYZZSWCNX-CBNWXSEKSA-L
C.I. 14010,5-(4-Sulfophenylazo)salicylic acid, disodium salt
9575878
disodium;(3E)-6-oxo-3-[(4-sulfonatophenyl)hydrazinylidene]cyclohexa-1,4-diene-1-carboxylate
C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
6054-99-5
C13H8N2Na2O6S
366.255
CWQLKNYZZSWCNX-CBNWXSEKSA-L
C.I. 14010,5-(4-Sulfophenylazo)salicylic acid, disodium salt
9575878
disodium;(3E)-6-oxo-3-[(4-sulfonatophenyl)hydrazinylidene]cyclohexa-1,4-diene-1-carboxylate
C1=CC(=CC=C1NN=C2C=CC(=O)C(=C2)C(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Dye Content 80% min.
Infrared Spectrum Authentic
Formula Weight 366.26
Physical Form Fine Crystalline Powder
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Yellow to Brown
Quantity 5g
Chemical Name or Material Mordant Yellow 10

GHS H Statement:
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-976-2

RTECSNumber : GB4450000

TSCA : TSCA