Learn More
Aviva Systems Biology Corp MORF4 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP47609_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MORF4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MORF4. Alternate names: CSR, CSRB, SEN, SEN1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59609
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.