Learn More
Morphix Technologies SafeAir™ Detection Badges
Single-level screening badges provide economical daily exposure monitoring.
Manufacturer:  Morphix Technologies 382000
 View more versions of this product
Catalog No. 18-999-1393
  Check Availability
Add to cart
Specifications
Description
When the target gas is detected at the threshold level, an immediate color change occurs and an exclamation point is visible within the color triangle on the badge.
  • Provide immediate results
  • Easy to read
  • No laboratory analysis is required
  • May be used for as little as 15 minutes or up to two days; for minimum/maximum recommended sampling times, exposure level, and minimum detectable limit for each analyte, contact your Fisher Safety Customer Service Representative
Order Info
Lightweight badges attach to user's collar with a clip (19-087-526, sold separately). Available 50 badges per pack.
Description & Specifications
Specifications
Phosgene
0.4ppm.Hour
55 to 99°F
0.003 lb.
Personnel or area monitoring for exposure times ranging from 15 min. to 24 Hours
Detection badge
12.77 to 37.22°C
0.001kg
Safety and Handling
ShelfLife : 1 Year
Accessories