Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morpholine, 99.5%, purified by redistillation, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
110-91-8
C4H9NO
87.12
YNAVUWVOSKDBBP-UHFFFAOYSA-N
1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro
8083
CHEBI:34856
morpholine
C1COCCN1
110-91-8
C4H9NO
87.12
YNAVUWVOSKDBBP-UHFFFAOYSA-N
1-oxa-4-azacyclohexane,tetrahydro-1,4-oxazine,diethylene oximide,diethylenimide oxide,diethyleneimide oxide,drewamine,diethylene imidoxide,tetrahydro-p-oxazine,tetrahydro-2h-1,4-oxazine,p-isoxazine, tetrahydro
8083
CHEBI:34856
morpholine
C1COCCN1
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.99
Refractive Index 1.4530 to 1.4550
Quantity 100mL
Density 0.9900g/mL
Viscosity 2.23 mPa.s (20°C)
Melting Point -5.0°C
Boiling Point 126.0°C to 130.0°C
Flash Point 32°C
Assay Percent Range 99.4% min. (GC)
Merck Index 15,6362
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, acetone, benzene,, ether, castor oil, ethylene glycol, 2-hexanone,, linseed oil, turpentine, pine oil
Formula Weight 87.12
Percent Purity 99.5%
Chemical Name or Material Morpholine
Show More Show Less

GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (

GHS Signal Word: Danger

EINECSNumber : 203-815-1