Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Morpholine, ≥99.5% (GC), MilliporeSigma™ Supelco™
Morpholines are systemic compounds that exert both protective and curative actions against fungal diseases.
$99.00
Specifications
CAS | 110-91-8 |
---|---|
Type | Analytical Standard |
Linear Formula | C4H9 NO |
MDL Number | MFCD00005972 |
Synonym | Tetrahydro-1,4-oxazine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-9649
|
MilliporeSigma Supelco
9017910MLF |
10 mL |
Each for $99.00
|
|
|||||
11-102-0336
|
MilliporeSigma Supelco
901795MLF |
5 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
110-91-8 | |
C4H9 NO | |
Tetrahydro-1,4-oxazine | |
≥99.5% (GC) | |
590°F | |
3 (vs. air) | |
-7°C to -5°C | |
31°C |
Analytical Standard | |
MFCD00005972 | |
87.12 | |
Analytical Standard | |
7 mmHg (20°C) | |
0.996 g/mL (at 25°C) | |
129°C | |
Neat |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title