Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Morpholinomethyl polystyrene resin, 1% DVB, 100-200 mesh, 2-3.5mmol/g, ACROS Organics
Manufacturer: Acros Organics 358370050
Specifications
crosslinking: 1% DVB | |
1% DVB, 2-3.5 mmol/g, 100-200 mesh | |
100 to 200 mesh | |
5g | |
MFCD00147659 |
Morpholinomethyl polystyrene resin | |
2 to 3.5mmol/g | |
Glass bottle | |
Cream to Yellow | |
Beads |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title