Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mosapride citrate, Tocris Bioscience™

Catalog Number 284410
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
112885-42-4
C27H33ClFN3O10
614.02
MFCD01666680
HUZTYZBFZKRPFG-UHFFFAOYNA-N
gasmot, mosapride citrate, +--4-amino-5-chloro-2-ethoxy-n-4-4-fluorobenzyl-2-morpholinyl methyl benzamide citrate, dsstox_cid_26207, dsstox_rid_81436, dsstox_gsid_46207, 4-amino-5-chloro-2-ethoxy-n-4-4-fluorobenzyl morpholin-2-yl methyl benzamide 2-hydroxypropane-1,2,3-tricarboxylate, benzamide, 4-amino-5-chloro-2-ethoxy-n-4-4-fluorophenyl methyl-2-morpholinyl methyl-, 2-hydroxy-1,2,3-propanetricarboxylate 1:1, citric acid; mosapride, 4-amino-5-chloro-2-ethoxy-n-4-4-fluorophenyl methyl-2-morpholinyl methyl-benzamide 2-hydroxy-1,2,3-propanetricarboxylate
119583
2-hydroxypropane-1,2,3-tricarboxylic acid; 4-amino-5-chloro-2-ethoxy-N-({4-[(4-fluorophenyl)methyl]morpholin-2-yl}methyl)benzamide
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCOC1=CC(N)=C(Cl)C=C1C(=O)NCC1CN(CC2=CC=C(F)C=C2)CCO1
This item is not returnable. View return policy
112885-42-4
C27H33ClFN3O10
614.02
MFCD01666680
HUZTYZBFZKRPFG-UHFFFAOYNA-N
gasmot, mosapride citrate, +--4-amino-5-chloro-2-ethoxy-n-4-4-fluorobenzyl-2-morpholinyl methyl benzamide citrate, dsstox_cid_26207, dsstox_rid_81436, dsstox_gsid_46207, 4-amino-5-chloro-2-ethoxy-n-4-4-fluorobenzyl morpholin-2-yl methyl benzamide 2-hydroxypropane-1,2,3-tricarboxylate, benzamide, 4-amino-5-chloro-2-ethoxy-n-4-4-fluorophenyl methyl-2-morpholinyl methyl-, 2-hydroxy-1,2,3-propanetricarboxylate 1:1, citric acid; mosapride, 4-amino-5-chloro-2-ethoxy-n-4-4-fluorophenyl methyl-2-morpholinyl methyl-benzamide 2-hydroxy-1,2,3-propanetricarboxylate
119583
2-hydroxypropane-1,2,3-tricarboxylic acid; 4-amino-5-chloro-2-ethoxy-N-({4-[(4-fluorophenyl)methyl]morpholin-2-yl}methyl)benzamide
OC(=O)CC(O)(CC(O)=O)C(O)=O.CCOC1=CC(N)=C(Cl)C=C1C(=O)NCC1CN(CC2=CC=C(F)C=C2)CCO1

5-HT4 agonist and 5-HT3 antagonist

5-HT4 receptor agonist and 5-HT3 receptor antagonist. Displays no activity at D2, α1, α2, 5-HT1 and 5-HT2 receptors. Gastroprokinetic agent; increases gastric emptying in rats and stimulates gastric motor actvity in conscious dogs.
Quantity 10 mg
Formula Weight 614.02
Percent Purity >99%
Chemical Name or Material Mosapride citrate
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit