Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOSC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550620

Catalog No. PA550620


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total MARC1 Antibody.

MOSC1 (MOCO sulphurase C-terminal domain containing 1), also known as MARC1, is a 337 amino acid mitochondrial protein that is thought to function as an oxidoreductase. Existing as three alternatively spliced isoforms, MOSC1 contains one MOSC domain and binds molybdenum as a cofactor. The gene encoding MOSC1 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MOSC1
Polyclonal
Unconjugated
MARC1
mARC1; mitochondrial amidoxime reducing component 1; mitochondrial amidoxime-reducing component 1; moco sulfurase C-terminal domain-containing protein 1; MOCO sulphurase C-terminal domain containing 1; molybdenum cofactor sulfurase C-terminal domain-containing protein 1; MOSC domain-containing protein 1; MOSC domain-containing protein 1, mitochondrial; MOSC1
Rabbit
Antigen affinity chromatography
RUO
64757
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
2.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q5VT66
MARC1
Synthetic peptide corresponding to a region derived from internal residues of human mitochondrial amidoxime reducing component 1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.