Learn More
Selleck Chemical LLCSupplier Diversity Partner Motesanib Diphosphate (AMG-706) 5mg 857876-30-3  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S10325MG
Motesanib Diphosphate (AMG-706) is a potent ATP-competitive inhibitor of VEGFR1/2/3 with IC50 of 2 nM/3 nM/6 nM, respectively; similar activity against Kit (c-Kit), ~10-fold more selective for VEGFR than PDGFR and Ret. Phase 3.
Catalog No. 50-621-0