Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform™ QUIK Sign Fold-Ups - SAFETY FIRST FOR THE SAFETY AND HEALTH OF OUR CUSTOMERS (Human With Arrows Symbol)
Accuform™ Floor Signs alert and remind everyone to practice social and physical distancing to prevent the spread of the Coronavirus (COVID-19)
Supplier: Accuform™ PFK900
Description
- Molded polyethylene plastic stand is 30.5 x 26 in. with feet 22 in. apart
- Includes one 18 x 24 in. corrugated plastic printed sign fits into slot
- Handle holes for easy portability
- Stand closes to 2 in. thick with molded in lug points for nesting into each other when stacked
Specifications
Floor Sign | |
Non-adhesive | |
SAFETY FIRST FOR THE SAFETY AND HEALTH OF OUR CUSTOMERS |
Plastic | |
30.5 x 26 in. | |
Assorted |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction