Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mott Manufacturing Altus™ Table System Component, Pencil Drawer

Mott Altus pencil drawer helps to keep workstation tidy and things within reach.

$415.50

Specifications

Type Drawer
For Use With (Equipment) Sigma Cart™ and Altus™ Tables
Width (Metric) 50.8 cm
View More Specs

Products 5
Catalog Number Mfr. No. Location Price Quantity & Availability  
Catalog Number Mfr. No. Location Price Quantity & Availability  
04-000-052
View Documents
Mott Manufacturing
PEN0020NE
Zone 4
Each for $415.50
Add to cart
 
04-000-139
View Documents
Mott Manufacturing
PEN0020MGC
Zone 7
Each for $415.50
Add to cart
 
04-000-574
View Documents
Mott Manufacturing
PEN0020SB
Zone 10
Each for $415.50
Add to cart
 
04-000-835
View Documents
Mott Manufacturing
PEN0020CC
Zone 1
Each for $415.50
Add to cart
 
04-000-226
View Documents
Mott Manufacturing
PEN0020DT
Zone 5
Each for $415.50
Add to cart
 
Description

Description

  • Suspends from tables with hanging rails
  • Supplied with full extension runners
Order Destinations
  • Zone 1 (Use CC suffix): Montana (Eastern), Minnesota, North Dakota, South Dakota, Wyoming
  • Zone 2 (Use WJ suffix): Iowa, Kansas, Missouri, Nebraska, Wisconsin
  • Zone 3 (Use SL suffix): Alabama, Florida, Georgia, Illinois, Kentucky, Tennessee
  • Zone 4 (Use NE suffix): Connecticut, Delaware, District of Columbia, Maine, Maryland, Massachusetts, New Hampshire, North Carolina, Rhode Island, South Carolina, Vermont, Virginia, West Virginia
  • Zone 5 (Previous DT suffix): Moved to Zone 9
  • Zone 6 (Previous CF suffix): Moved to Zone 9
  • Zone 7 (Use MGC suffix): Arizona, Arkansas, Colorado, Louisiana, Mississippi, New Mexico, Oklahoma, Texas
  • Zone 8 (Use S suffix): New Jersey, New York, Pennsylvania (Eastern)
  • Zone 9 (Use SV suffix): Michigan, New York State (Upper), Ohio, Pennsylvania (Western)
  • Zone 10 (Use SB suffix): Alaska, California, Hawaii, Idaho, Montana (Western), Nevada, Oregon, Utah, Washington

Order Info

When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to

Specifications

Specifications

Drawer
Sigma Cart™ and Altus™ Tables
50.8 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit