Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mott Manufacturing Altus™ Table System Component, Steel Top Shelf Assembly
SDP

Mott steel top shelf assembly adds more room for storage on Altus workbench.

$159.00 - $187.30

Specifications

Product Type Shelf Assembly
Color Gray
Material Steel
View More Specs

Products 15
Catalog Number Mfr. No. Location Width (English) Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Location Width (English) Width (Metric) Price Quantity & Availability  
04-000-028
SDP
View Documents
Mott Manufacturing
AAS1248NE
Zone 4 48 in. 121.9 cm
Each for $159.00
Add to Cart
 
04-000-029
SDP
View Documents
Mott Manufacturing
AAS1260NE
Zone 4 60 in. 152.4 cm
Each for $176.40
Add to Cart
 
04-000-030
SDP
View Documents
Mott Manufacturing
AAS1272NE
Zone 4 72 in. 182.8 cm
Each for $187.30
Add to Cart
 
04-000-115
SDP
View Documents
Mott Manufacturing
AAS1248MGC
Zone 7 48 in. 121.9 cm
Each for $159.00
Add to Cart
 
04-000-116
SDP
View Documents
Mott Manufacturing
AAS1260MGC
Zone 7 60 in. 152.4 cm
Each for $176.40
Add to Cart
 
04-000-117
SDP
View Documents
Mott Manufacturing
AAS1272MGC
Zone 7 72 in. 182.8 cm
Each for $187.30
Add to Cart
 
04-000-551
View Documents
Mott Manufacturing
AAS1260SB
Zone 10 60 in. 152.4 cm
Each for $176.40
Add to Cart
 
04-000-550
View Documents
Mott Manufacturing
AAS1248SB
Zone 10 48 in. 121.9 cm
Each for $159.00
Add to Cart
 
04-000-552
View Documents
Mott Manufacturing
AAS1272SB
Zone 10 72 in. 182.8 cm
Each for $187.30
Add to Cart
 
04-000-811
View Documents
Mott Manufacturing
AAS1248CC
Zone 1 48 in. 121.9 cm
Each for $159.00
Add to Cart
 
04-000-812
View Documents
Mott Manufacturing
AAS1260CC
Zone 1 60 in. 152.4 cm
Each for $176.40
Add to Cart
 
04-000-813
View Documents
Mott Manufacturing
AAS1272CC
Zone 1 72 in. 182.8 cm
Each for $187.30
Add to Cart
 
04-000-202
View Documents
Mott Manufacturing
AAS1248DT
Zone 2 48 in. 121.9 cm
Each for $159.00
Add to Cart
 
04-000-203
View Documents
Mott Manufacturing
AAS1260DT
Zone 5 60 in. 152.4 cm
Each for $176.40
Add to Cart
 
04-000-204
View Documents
Mott Manufacturing
AAS1272DT
Zone 2 72 in. 182.8 cm
Each for $187.30
Add to Cart
 
Description

Description

  • Includes 2.54cm (1 in.) thick, 30.4cm (12 in.) deep shelf, pair of 30cm (12 in.) deep brackets
  • Shelves are adjustable in 2.54cm (1 in.) increments
  • For use with Altus products only
Order Destinations
  • Zone 1 (Use CC suffix): Montana (Eastern), Minnesota, North Dakota, South Dakota, Wyoming
  • Zone 2 (Use WJ suffix): Iowa, Kansas, Missouri, Nebraska, Wisconsin
  • Zone 3 (Use SL suffix): Alabama, Florida, Georgia, Illinois, Kentucky, Tennessee
  • Zone 4 (Use NE suffix): Connecticut, Delaware, District of Columbia, Maine, Maryland, Massachusetts, New Hampshire, North Carolina, Rhode Island, South Carolina, Vermont, Virginia, West Virginia
  • Zone 5 (Previous DT suffix): Moved to Zone 9
  • Zone 6 (Previous CF suffix): Moved to Zone 9
  • Zone 7 (Use MGC suffix): Arizona, Arkansas, Colorado, Louisiana, Mississippi, New Mexico, Oklahoma, Texas
  • Zone 8 (Use S suffix): New Jersey, New York, Pennsylvania (Eastern)
  • Zone 9 (Use SV suffix): Michigan, New York State (Upper), Ohio, Pennsylvania (Western)
  • Zone 10 (Use SB suffix): Alaska, California, Hawaii, Idaho, Montana (Western), Nevada, Oregon, Utah, Washington

Order Info

When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to

Specifications

Specifications

Shelf Assembly
Gray
Steel
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit