Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Mott Manufacturing RFV2™ Vertical Sash Fume Hood Accessory, Epoxy Worktop
Use a Mott Epoxy Worktop to protect your fume hood from damage. These come with cupsinks that on the right and designed for 18 in. deep fume hood base cabinets.
Manufacturer:  Mott Manufacturing RFR2672S
 View more versions of this product
Catalog No. 01-100--206
Description
Description
  • Black epoxy work surface with cupsink on right side for RFV2 Series fume hoods
  • Cupsinks are located for 18 in. deep fume hood base cabinets (not included)
Order Info
When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to.
Specifications
Specifications
Epoxy
RFV2™ Vertical Sash Fume Hood
66.04 x 182.88 x 2.54cm (26 x 72 x 1 in.)
Right-side Cupsink
Black
Zone 5