Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mott Manufacturing RFV2™ Vertical Sash Fume Hood Accessory, Epoxy Worktop
Use a Mott Epoxy Worktop to protect your fume hood from damage. These come with cupsinks that on the right and designed for 18 in. deep fume hood base cabinets.
Manufacturer: Mott Manufacturing RFR2648NE
Description
- Black epoxy work surface with cupsink on right side for RFV2 Series fume hoods
- Cupsinks are located for 18 in. deep fume hood base cabinets (not included)
Order Info
When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to.
Specifications
Epoxy | |
RFV2™ Vertical Sash Fume Hood | |
66.04 x 121.92 x 2.54cm (26 x 48 x 1 in.) |
Right-side Cupsink | |
Black | |
Zone 4 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title