Mott Manufacturing Steel Casework Suspended Cabinet, Drawer Units

Suspended cabinet with three drawers for maximum storage

Manufacturer: Mott ManufacturingSupplier Diversity Partner 1950044DT

 View more versions of this product

Catalog No. 24-200-296

  Check Availability
Add to Cart

    Specifications

    Warranty Two years form the date of shipment
    Certifications SEFA, ISO9001:2008
    For Use With (Equipment) Steel Casework Suspended Cabinet
    Color Gray
    No. of Drawers 3
    Location Zone 5
    Dimensions (D x W x H) 55.8 x 38.1 x 47.6cm (22 x 15 x 18.75 in.)
    Product Type Drawer Unit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit