Learn More
Novus Biologicals™ Mouse ABHD6 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP266380
Description
The Mouse ABHD6 ELISA Kit (Colorimetric) measures mouse ABHD6 in serum, plasma and other biological fluids.Specifications
31.25 - 2000 pg/mL | |
This kit recognizes Mouse ABHD6 in samples. No significant cross-reactivity or interference between Mouse ABHD6 and analogues was observed. | |
57406 | |
Mouse | |
1 Kit | |
Storage is content dependent. |
18.75 pg/mL | |
ELISA | |
ABHD6 | |
ELISA Kit (Colorimetric) | |
Biological Fluids,Plasma,Serum | |
ABHD6 |