Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Adipoq (NP_033735, 111 a.a. - 247 a.a.) Partial Recombinant Protein

Used for SDS-PAGE

Manufacturer:  Abnova™ P4326

Catalog No. 89-932-710


Add to cart

Description

Description

Sequence: MAYMYRSAFSVGLETRVTVPNVPIRFTKIFYNQQNHYDGSTGKFYCNIPGLYYFSYHITVYMKDVKVSLFKKDKAVLFTYDQYQEKNVDQASGSVLLHLEVGDQVWLQVYGDGDHNGLYADNVNDSTFTGFLLYHDTN
Specifications

Specifications

Escherichia coli expression system
1 mg/mL
Liquid
11450
Adipoq (Mouse) Recombinant Protein
Conventional Chromatography
15% SDS-PAGE Stained with Coomassie Blue
MAYMYRSAFSVGLETRVTVPNVPIRFTKIFYNQQNHYDGSTGKFYCNIPGLYYFSYHITVYMKDVKVSLFKKDKAVLFTYDQYQEKNVDQASGSVLLHLEVGDQVWLQVYGDGDHNGLYADNVNDSTFTGFLLYHDTN
RUO
Adipoq
Escherichia coli
None
NP_033735
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
16kDa
Escherichia coli expression system
>95% by SDS-PAGE
100 ug
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
30kDa/APN/Acdc/Acrp30/GBP28/adipo/apM1
Adipoq
Yes
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit