Learn More
Abnova™ Mouse Adipoq (Q60994, 145 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4528
Catalog No. 89-932-712
Add to cart
Description
Description
Specifications
Specifications
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
16.6kDa
Escherichia coli expression system
25 ug
RUO
30kDa/APN/Acdc/Acrp30/GBP28/adipo/apM1
Adipoq
Escherichia coli
None
Q60994
Lyophilized
11450
Adipoq (Mouse) Recombinant Protein
≥98% by SDS-PAGE
Store at -20°C on dry atmosphere. After reconstitution with 5 mM Tris, pH 7.6, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Adipoq
Determined by its ability to inhibit the proliferation of murine myeloid cell lines M1. The ED50 for this effect is less than 15μg/mL.
Yes