Learn More
Abnova™ Mouse Adipoq (Q60994) Full-length Recombinant Protein
Used for SDS-PAGE
Manufacturer:  Abnova™ P4530
Catalog No. 89-932-713
Add to cart
Description
Description
Specifications
Specifications
Q60994
Lyophilized
11450
Adipoq (Mouse) Recombinant Protein
25 ug
RUO
30kDa/APN/Acdc/Acrp30/GBP28/adipo/apM1
Adipoq
Yes
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized from 25mM Tris, 0.15M NaCl, pH 7.4 (1.250mg BSA)
30kDa
≥90% by SDS-PAGE
Store at -20°C on dry atmosphere. After reconstitution with PBS containing at least 0.1% BSA, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Adipoq
Escherichia coli
None