Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse AgRP/ART Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF634
This item is not returnable.
View return policy
Description
AgRP/ART Polyclonal specifically detects AgRP/ART in Mouse samples. It is validated for Immunohistochemistry.Specifications
AgRP/ART | |
Polyclonal | |
Unconjugated | |
P56473 | |
AGRP | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse AgRP/ART Val25-Thr131 Accession # P56473 | |
100 μg | |
Primary | |
Detects mouse AgRP in direct ELISAs. In direct ELISAs, less than 25% cross-reactivity with recombinant human AgRP is observed. | |
Mouse | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 1.7-5 ug/mL | |
agouti related protein homolog (mouse), agouti-related protein, Agrt, AGRTMGC118963, ART, ARTagouti (mouse) related protein, ASIP2 | |
Goat | |
Affinity Purified | |
RUO | |
181 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction