Learn More
Novus Biologicals™ Mouse Alpha Actinin 2 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP266418
Catalog No. NBP266418
Add to Cart
Description
Description
The Mouse Alpha Actinin 2 ELISA Kit (Colorimetric) measures mouse Alpha Actinin 2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Mouse ACTN2 in samples. No significant cross-reactivity or interference between Mouse ACTN2 and analogues was observed.
Biological Fluids, Plasma, Serum
88
Mouse
Storage of components varies. See protocol for specific instructions.
0.19 ng/mL
ELISA Kit (Colorimetric)
ELISA
ACTN2
1 Kit
Alpha Actinin 2
Product Suggestions
Product Suggestions