Learn More
Novus Biologicals™ Mouse alpha-Smooth Muscle Actin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266429
Catalog No. NBP266429
Add to cart
Description
Description
The Mouse alpha-Smooth Muscle Actin ELISA Kit (Colorimetric) measures mouse alpha-Smooth Muscle Actin in serum, plasma and other biological fluids.
Specifications
Specifications
3.13 - 200 ng/mL
This kit recognizes Rat ACTalpha2 in samples. No significant cross-reactivity or interference between Mouse ACTalpha2 and analogues was observed.
59
Mouse
1 Kit
Biological Fluids,Plasma,Serum
Storage is content dependent.
alpha-Smooth Muscle Actin
1.88 ng/mL
ELISA
ACTA2
ELISA Kit (Colorimetric)
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
This kit recognizes mouse ACTalpha2 in samples. No significant cross-reactivity or interference between Mouse ACTalpha2 and analogues was observed.
AAT6, actin, alpha 2, smooth muscle, aorta, actin, aortic smooth muscle, ACTSA, ACTVS, alpha 2 actin, alpha SMA, alpha smooth muscle actin, alpha-actin-2, alpha-SMA, a-sma, Cell growth-inhibiting gene 46 protein, growth-inhibiting gene 46, sma, smooth muscle actin