Learn More
Novus Biologicals™ Mouse Amylin ELISA Kit (Colorimetric)
Mouse Amylin ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: Novus Biologicals™ NBP276734
Description
The Mouse Amylin ELISA Kit (Colorimetric) measures mouse Amylin in serum, plasma and other biological fluids.Specifications
62.5 to 4000 pg/mL | |
ELISA Kit (Colorimetric) | |
3375 | |
Mouse | |
Storage of components varies. See protocol for specific instructions. |
37.5 pg/mL | |
Biological Fluids, Plasma, Serum | |
IAPP | |
1 Kit |