Learn More
3-DG-imidazolone, Mouse, Clone: JNH 27, Abnova™
Mouse monoclonal antibody raised against 3-DG-imidazolone.
Manufacturer:  Abnova Corporation MAB1738
Catalog No. 89-135-040
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
3-DG-imidazolone
JNH 27
Mouse monoclonal antibody raised against 3-DG-imidazolone.
In PBS (0.1% proclin, 2.0% Block Ace)
Mouse
IgG1
RUO
Primary
Immunohistochemistry
Unconjugated
Immunohistochemistry (7 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
3-DG-imidazolone conjugated with HSA.
50 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal