Learn More
Mouse anti-ABCG2/CD338, Clone: MM0047-2J39, Alexa Fluor 488, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11093511AF488
Catalog No. NB11093511X
Add to cart
Description
Description
ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
ABCG2/CD338
MM0047-2J39
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
ABCG2
human recombinant rHABCG2 protein
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This was selected for its ability to detect human ABCG2 by flow cytometry and immunocytochemistry.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Mouse
IgG2
0.1 ml
ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cells
Primary
9429
Human