Learn More
Adiponectin, Mouse, Clone: Adn305, Abnova™
Mouse monoclonal antibody raised against native adiponectin.
Manufacturer:  Abnova Corporation MAB8471
Catalog No. 89-136-547
Add to cart
Description
Description
Specifications
Specifications
Adiponectin
Adn305
Mouse monoclonal antibody raised against native adiponectin.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG1
RUO
Primary
Recommended pairs for total adiponectin immunodetection in sandwich immunoassay (capture-detection) : Adn305-Adn279.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native purified adiponectin from human serum.
1 mg
Store at 4°C.
Monoclonal
Human