Learn More
AFP, Mouse, Clone: AP-1, Abnova™
Mouse monoclonal antibody raised against native AFP.
Supplier: Abnova Corporation MAB1508
Description
This gene encodes alpha-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. [provided by RefSeq
Specifications
AFP | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
AFP | |
AFP | |
Native purified human AFP. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Immunohistochemistry (PFA fixed) | |
C3 | |
Mouse monoclonal antibody raised against native AFP. | |
Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
FETA/HPAFP | |
Mouse | |
Affinity purification | |
RUO | |
174 | |
Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.