Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Agrin, Clone: Agr 33, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB120123580.2MG
Description
Agrin Monoclonal specifically detects Agrin in Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Agrin | |
Monoclonal | |
1.0 mg/mL | |
Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
agrin, agrin proteoglycan | |
Mouse | |
Protein G purified | |
RUO | |
375790 | |
Mouse, Rat | |
Purified |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Agr 33 | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
AGRN | |
Recombinant rat agrin, C-terminal construct. | |
0.2 mg | |
Primary | |
detects all isoforms of agrin. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction