Learn More
Mouse anti-AKT, Clone: MM0647-2L45, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236780
Catalog No. NBP236780
Add to cart
Description
Description
AKT Monoclonal antibody specifically detects AKT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Specifications
Specifications
AKT
MM0647-2L45
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with recombinant human AKT1).
Protein G purified
RUO
Primary
207
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
LYOPH
Western Blot 1:1000-2000, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:50-200, CyTOF-ready
P31749
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG2
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human AKT1, 2 and 3 by WB. Cross reacts with mouse and rat AKTs proteins.
Human