Learn More
Mouse anti-AKT (p Thr308), Clone: MM0650-9F42, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236782
Catalog No. NBP236782
Add to cart
Description
Description
AKT p Thr308 Monoclonal antibody specifically detects AKT p Thr308 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
AKT (p Thr308)
MM0650-9F42
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
AKT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing the human Akt1 T308 site.
Protein G purified
RUO
Primary
207
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Immunocytochemistry, Immunofluorescence, Western Blot
LYOPH
Western Blot 1:1000-2000, Immunocytochemistry/Immunofluorescence 1:50-200
P31749
RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB-ALPHA, PRKBA, RAC
Mouse
IgG3
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognizes human AKT1 with phospho-T308.
Human