Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-alpha-Fetoprotein/AFP, Clone: SPM334, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2377540.1MG
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human, Porcine, Canine, Monkey, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
alpha-Fetoprotein/AFP | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml, Immunohistochemistry 0.5-1ug/ml, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml, CyTOF-ready | |
P02771 | |
AFP | |
Alpha fetoprotein (AFP) purified from serum of a hepatoma patient | |
0.1 mg | |
Primary | |
Antibody recognizes an oncofetal glycoprotein with a single chain of 70kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. Does not react with Bovine, Feline, Mouse or Rat. | |
Human, Porcine, Canine, Primate, Bovine (Negative), Feline (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
SPM334 | |
Unconjugated | |
10mM PBS - BSA free with No Preservative | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
Mouse | |
Protein A purified | |
RUO | |
174 | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction