Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Androgen R/NR3C4, Clone: SPM335, Novus Biologicals™

Mouse Monoclonal Antibody

$129.00 - $299.00

Specifications

Antigen Androgen R/NR3C4
Clone SPM335
Concentration 0.2mg/mL
Immunogen A synthetic peptide, aa 299-315, (STEDTAEYSPFKGGYTK) of human AR.
Gene ID (Entrez) 367
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24478701 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2447870.1MG
0.1 mg Each for $299.00
Add to cart
 
NBP24478700 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2447870.02MG
0.02 mg Each for $129.00
Add to cart
 
NBP24478702 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2447870.2MG
0.2 mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human, Mouse (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

Androgen R/NR3C4
0.2mg/mL
367
Monoclonal
Mouse
IgG1 Kappa
Purified
RUO
Protein A or G purified
SPM335
A synthetic peptide, aa 299-315, (STEDTAEYSPFKGGYTK) of human AR.
1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide
Primary
Recognizes a protein of 110kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. It does not cross react with estrogen, progesterone, or glucocorticoid receptors. The expression of AR is reportedly inversely correlated with histologic grade i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed, as a marker of hormone-responsiveness and thus it may be useful in identifying patients likely to benefit from anti-androgen therapy. Anti-androgen receptor has been useful clinically in differentiating morpheaform basal cell carcinoma (mBCC) from desmoplastic trichoepithelioma (DTE) in the skin.This MAb is superb for staining of formalin/paraffin tissues.
Unconjugated
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Store at 4C.

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit