Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-APAF-1, Clone: 5E11, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241323

Catalog No. NBP241323

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

APAF-1
500000000000
Unconjugated
PBS with 0.02% Sodium Azide
APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413
Mouse
IgG2a/IgG2b
0.1 mg
Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors
Primary
317
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
1 mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500, Immunocytochemistry
Affinity Purified
APAF1
A peptide corresponding to amino acids near the carboxy terminus of human Apaf1.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit