Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-APC-PCI Complex, Clone: M36, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2413800.2MG

Catalog No. NBP24138002


Add to cart

Description

Description

Ensure accurate, reproducible results in Enzyme Linked Immunosorbent Assay
Specifications

Specifications

APC-PCI Complex
M36
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Mouse
IgG1 Kappa
200 ug
Store at 4C in the dark.
Monoclonal
Human
ELISA
1 mg/mL
ELISA 1:100-1:2000
Purified
Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%).
Protein A or G purified
RUO
Primary
Antibody is specific for APC:PCI complex and cleaved PCI but has little reactivity towards native PCI. The dissociation constants (as determined by standard surface plasmon resonance technique) are: Kd= 4x10e-10 M for PCI in complex with APC and 2x10e-10 M for cleaved PCI. The value of Kd for native PCI is too low to measure, i.e. > 10e-5 M.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only