Learn More
Mouse anti-Apolipoprotein A5, Clone: 4H8H8E2, Alexa Fluor 700, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11057307AF700
Catalog No. NB11057307Z
Add to cart
Description
Description
Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Apolipoprotein A5
4H8H8E2
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Q6Q788
Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3
Mouse
IgG1 Kappa
0.1 ml
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
116519
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Purified
APOA5
Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProt# Q6Q788]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human