Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGDIB, Mouse, FITC, Clone: 97A1015, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of ARHGDIB.
Supplier: Abnova Corporation MAB0060
Specifications
ARHGDIB | |
Monoclonal | |
FITC | |
The optimal working dilution should be determined by the end user. | |
ARHGDIB | |
ARHGDIB | |
A synthetic peptide corresponding to the region of the Fas-induced cleavage site of human ARHGDIB. | |
RUO | |
397 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |
Flow Cytometry | |
97A1015 | |
Mouse monoclonal antibody raised against synthetic peptide of ARHGDIB. | |
In PBS (0.05% sodium azide) | |
D4/GDIA2/GDID4/LYGDI/Ly-GDI/RAP1GN1/RhoGDI2 | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction