Learn More
Mouse anti-ATG12, Clone: 9411, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243753
Catalog No. NBP243753
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
ATG12 Monoclonal antibody specifically detects ATG12 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
ATG12
9411
Western Blot 1:500-1:3000
Purified
ATG12
The exact sequence is proprietary.
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Western Blot
Unconjugated
PBS with No Preservative
Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog
Mouse
IgG1
0.1 ml
Apoptosis, Autophagy, Neuroscience
Primary
9140