Learn More
Mouse anti-B7-1/CD80, Clone: 2D10.4, PE-Cy5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP200123100 TESTS
 View more versions of this product
Catalog No. NBP200123
Add to cart
Description
Description
B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
B7-1/CD80
2D10.4
Flow Cytometry 1:10-1:1000
Affinity Purified
CD80
The immunogen for this antibody was CD80.
Protein A or G purified
RUO
Primary
941
Flow Cytometry
PE-Cyanine5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Mouse
IgG1 Kappa
100 Tests
Store at 4C. Do not freeze.
Monoclonal
Human