Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-B7-1/CD80, Clone: 2D10.4, PE-Cy5, Novus Biologicals™

Mouse Monoclonal Antibody

$180.00 - $419.00

Specifications

Antigen B7-1/CD80
Clone 2D10.4
Immunogen The immunogen for this antibody was CD80.
Gene ID (Entrez) 941
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP20012325
View Documents
Novus Biologicals
NBP20012325 TESTS
25 Tests
Each for $180.00
Add to cart
 
NBP200123
View Documents
Novus Biologicals
NBP200123100 TESTS
100 Tests
Each for $419.00
Add to cart
 
Description

Description

B7-1/CD80 Monoclonal antibody specifically detects B7-1/CD80 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

B7-1/CD80
The immunogen for this antibody was CD80.
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Primary
Mouse
PE-Cyanine5
Protein A or G purified
Flow Cytometry
2D10.4
941
Monoclonal
Store at 4C. Do not freeze.
IgG1 Kappa
Affinity Purified
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit