Learn More
Mouse anti-B7-1/CD80, Clone: mAb104, Novus Biologicals™
Manufacturer:  Novus Biologicals DDX0160P1000.05MG
Catalog No. DDX0160P05
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
B7-1/CD80
mAb104
Unconjugated
Tris-NaCl (pH 8.0) with No Preservative
Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Mouse
IgG1
0.05 mg
Adaptive Immunity, Cytokine Research, Immunology, Signal Transduction
Primary
941
Flow Cytometry, Functional Assay
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Functional
Purified
CD80
CD80 was used as immunogen.
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human