Learn More
Mouse anti-Basophils, Clone: 212H6.16, Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0030B1000.05MG
Catalog No. DDX0030B05
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Basophils Monoclonal antibody specifically detects Basophils in Human, Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Basophils
212H6.16
Biotin
PBS and 50% Glycerol with No Preservative
Purified
Mouse
IgM
0.05 mg
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human, Canine
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
NP_006857
Anti-Basophils, Basophil
Basophil was used as immunogen.
Ion exchange chromatography
RUO
Primary
No-cross reactivity on mouse basophil