Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoylecgonine, Mouse, Clone: 2C4D5, Abnova™

Mouse monoclonal antibody raised against Benzoylecgonine.

Manufacturer:  Abnova Corporation MAB1541

Catalog No. 89-134-990


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Specifications

Specifications

Benzoylecgonine
2C4D5
Mouse monoclonal antibody raised against Benzoylecgonine.
In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Mouse
IgG1
RUO
Primary
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Benzoylecgonine conjugated with BSA.
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit