Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-beta 2-Microglobulin, Clone: SPM374, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2444660.02MG

 View more versions of this product

Catalog No. NBP24446600


Add to cart

Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence

beta 2-Microglobulin Monoclonal antibody specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence.
Specifications

Specifications

beta 2-Microglobulin
SPM374
Unconjugated
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Purified
B2M
Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient.
12 kDa
0.02 mg
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
Primary
567
Human, Primate (Negative)
Flow Cytometry, Immunofluorescence
0.2 mg/mL
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml
P61769
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
Mouse
IgG2a Kappa
Protein A or G purified
RUO
Store at 4C.
Monoclonal
Recognizes a protein of 12kDa, identified as b2-microglobulin. Beta-2-microglobulin non-covalently associates with the 44kDa a chain to form the HLA Class I antigen complex. Human beta-2-microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human b2-microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of beta2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only