Learn More
Beta-Galactosidase, Mouse, Clone: DC3 10D6, Abnova™
Mouse monoclonal antibody raised against Beta-Galactosidase.
Manufacturer:  Abnova Corporation MAB7236
Catalog No. 89-136-325
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Beta-Galactosidase
DC3 10D6
Mouse monoclonal antibody raised against Beta-Galactosidase.
In PBS
Mouse
IgG1
RUO
Primary
beta Galactosidase (DC3 10D6).
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Beta-Galactosidase from Escherichia coli.
100 ug
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal