Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BKbeta3a, Mouse, Clone: S40B-18, Abnova™

Mouse monoclonal antibody raised against partial recombinant BKbeta3a.

Manufacturer:  Abnova Corporation MAB7893

Catalog No. 89-136-395


Add to cart

Description

Description

Specifications

Specifications

BKbeta3a
S40B-18
Mouse monoclonal antibody raised against partial recombinant BKbeta3a.
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Mouse
IgG1
RUO
Primary
No cross-reactivity against BKbeta1, BKbeta1, BKbeta3b or BKbeta4.
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Western Blot (1-10 ug/mL) Immunohistochemistry (0.1-1.0 ug/mL) Immunocytochemistry (0.1-1.0 ug/mL) Immunofluorescence (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to N-terminus amino acids 1-49 of mouse BKbeta3a.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit