Learn More
Blood Group A Mouse anti-Human, Brilliant Violet 421, Clone: NaM87-1F6, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565386
Catalog No. BDB565386
Add to cart
Description
Description
The NaM87-1F6 monoclonal antibody specifically binds to blood group A expressed on human erythrocytes. Clone NaM87-1F6 induces agglutination of human group A red blood cells. It does not react with human group B, or group O red blood cells.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
Specifications
Blood Group A
NaM87-1F6
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Brilliant Violet 421
Affinity Purified
IgG3, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal